Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002046SS
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).Specifications
IgG1 Fc | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein A purified | |
RUO | |
Subclass-specific for human IgG1. No allotype restriction. Fc - region specific. | |
Store at 4C. Do not freeze. | |
IgG1 |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
2C11 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum. | |
0.04 mg | |
Secondary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction