Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc), Novus Biologicals™

Mouse Monoclonal Antibody
$169.20
Specifications
Antigen | IgG1 Fc |
---|---|
Clone | 2C11 |
Dilution | ELISA, Immunohistochemistry, Immunohistochemistry-Frozen |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1002046
![]() |
Novus Biologicals
NB1002046 |
0.2 mg |
Each for $169.20
|
|
|||||
NB1002046SS
![]() |
Novus Biologicals
NB1002046SS |
0.04 mg | N/A | N/A | N/A | ||||
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).Specifications
IgG1 Fc | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
Unconjugated | |
Mouse | |
Human | |
IgG1 | |
Protein A purified | |
Subclass-specific for human IgG1. No allotype restriction. Fc - region specific. |
2C11 | |
Monoclonal | |
Purified | |
RUO | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum. | |
Secondary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title