Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human IgG2 Fc Secondary Antibody (3C7), Novus Biologicals™
SDP

Catalog No. NB002825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002825 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002825 Supplier Novus Biologicals Supplier No. NBP268315500UG

Mouse Monoclonal Antibody

IgG2 Fc Monoclonal specifically detects IgG2 Fc in Human samples. It is validated for ELISA.

Specifications

Antigen IgG2
Applications ELISA
Classification Monoclonal
Clone 3C7
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Gene Alias Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker)
Gene Symbols IGHG2
Host Species Mouse
Immunogen Human IgG from pooled serum
Quantity 500 μg
Gene ID (Entrez) 3501
Test Specificity Mouse anti Human IgG2 antibody, clone 3C7, is specific for human IgG2 and recognizes an epitope in the Fc region.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.