Learn More
IgM Mouse anti-Human, Brilliant Violet 786, Clone: G20-127, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740998
Description
IgM is an important component in the first line of defense against foreign pathogens, but may also play a role in autoimmune diseases. IgM monomers consist of two light and two heavy chains. Unlike the heavy chain of an IgG antibody which contains 3 constant Ig domains, the µ heavy chain of IgM contains 4 constant Ig domains. Five IgM monomers complex with a small polypeptide (J-chain) to form pentameric IgM that can be found in human plasma. In an immune response, the binding of IgM to a cell surface antigen enables C1q to activate interactions with downstream components in the classical complement pathway. Mature B lymphocytes express IgM. The G20-127 monoclonal antibody binds to the heavy chain of human IgM. The G20-127 antibody is not thought to react with other immunoglobulin heavy chain isotypes.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
| IgM | |
| Monoclonal | |
| 0.2mg/mL | |
| IGHM; MU; Ig mu chain C region; AGM1; VH | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G20-127 | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only