Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Human IgM Fc Secondary Antibody (CH2), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500468APC
Description
IgM Fc Monoclonal antibody specifically detects IgM Fc in Human samples. It is validated for Flow Cytometry.Specifications
IgM Fc | |
Monoclonal | |
APC | |
Mouse | |
Ion exchange chromatography | |
RUO | |
The antibody CH2 reacts with Fc fragment of human IgM. | |
Store at 4C in the dark. |
Flow Cytometry | |
CH2 | |
Flow Cytometry | |
Purified human IgM. | |
0.1 mL | |
Secondary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction