Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human immunodeficiency virus reverse transcriptase, Mouse, Clone: AS34, Abnova™

Catalog No. 89135062 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-062 Supplier Abnova Corporation Supplier No. MAB1894

Mouse monoclonal antibody raised against recombinant human immunodeficiency virus reverse transcriptase.

Specifications

Antigen Human immunodeficiency virus reverse transcriptase
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone AS34
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human immunodeficiency virus reverse transcriptase.
Dilution ELISA (200-500 ng/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (without Mg2+ and Ca2+) (0.08% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to human immunodeficiency virus reverse transcriptase.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes an epitope on HIV-RT that is different from AS35-P epitope.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.