Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human immunodeficiency virus type 1 gp120, Mouse, Clone: G2-25, Abnova™

Catalog No. 89136237 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
125 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-237 125 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-237 Supplier Abnova Corporation Supplier No. MAB6566

Mouse monoclonal antibody raised against synthetic peptide of human immunodeficiency virus type 1 gp120.

Specifications

Antigen Human immunodeficiency virus type 1 gp120
Applications Inhibition Assays
Classification Monoclonal
Clone G2-25
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human immunodeficiency virus type 1 gp120.
Dilution Inhibition Assay (50 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Host Species Mouse
Immunogen A synthetic peptide corresponding to gp120 region of NL4-3 human immunodeficiency virus type 1 strain.
Quantity 125 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.