Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human immunodeficiency virus type 1 P24 (core protein), Mouse, Clone: 1027, Abnova™

Catalog No. 89136184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-184 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-136-184 Supplier Abnova Corporation Supplier No. MAB6350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 P24 (core protein).

Specifications

Antigen Human immunodeficiency virus type 1 P24 (core protein)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1027
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 P24 (core protein).
Dilution Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Host Species Mouse
Immunogen Recombinant protein corresponding to human immunodeficiency virus type 1 P24 core protein.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with HIV-1 P24 (core protein).
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.