Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human immunodeficiency virus type 1 Tat, Clone: 15 (02-003), Abnova™

Catalog No. 89136372 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-372 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-372 Supplier Abnova Corporation Supplier No. MAB7844
Add to Cart
Add to Cart

Specifications

Applications ELISA, Western Blot
Classification Monoclonal
Clone 15
Description Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 Tat.
Dilution ELISA (0.2-1 ug/mL) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to human immunodeficiency virus type 1 (BH10) Tat.
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Mapped to amino acids 2-9 (EPVDPRLE - B subtype a.a. consensus). This antibody recognizes the rTat of HIV-1 subtypes A (DPVDPNLE), B (EPVDPRLE) and C (EPVDPNLE). It reacts with HIV-1 Tat of BH10 and IIIB strains.
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.