Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human papilloma virus type 16 E1+E4, Mouse, Clone: TVG 403, Abnova™

Catalog No. 89136303 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-303 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-303 Supplier Abnova Corporation Supplier No. MAB7005
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human papilloma virus type 16 E1+E4.

Specifications

Antigen Human papilloma virus type 16 E1+E4
Applications ELISA, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone TVG 403
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human papilloma virus type 16 E1+E4.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Recombinant beta-galactosidase fusion protein corresponding E1/E4 polypeptide amino acids 36-41 of human papilloma virus type 16.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HPV16 E1+E4 (TVG 403). E1/E4 protein is aa 36-41 of human papilloma virus type 16.
Target Species Human Papillomavirus (HPV)
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.