Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-As, Mouse, Clone: IA-G1, Abnova™
Mouse monoclonal antibody raised against native I-As.
Supplier: Abnova Corporation MAB1334
Specifications
I-As | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
200 μg | |
Primary | |
Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b |
Flow Cytometry, Immunoprecipitation | |
IA-G1 | |
Mouse monoclonal antibody raised against native I-As. | |
Lyophilized from PBS | |
Native purified from mouse lymph node B cells. | |
RUO | |
React with mouse I-As. Cross react with I-Ap, not cross react with I-Ad, I-Ab, I-Ak. | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction