Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borrelia burgdorferi, Mouse, Clone: 5015, Abnova™

Catalog No. 89135536 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-536 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-536 Supplier Abnova Corporation Supplier No. MAB3899
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Borrelia burgdorferi.

Specifications

Antigen Borrelia burgdorferi
Applications ELISA
Classification Monoclonal
Clone 5015
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Borrelia burgdorferi.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native from Borrelia burgdorferi.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the 28KDa (Osp A) protein of Borrelia burgdorferi.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.