Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia species LPS, Mouse, Clone: BDI168, Abnova™

Catalog No. 89135552 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-552 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-552 Supplier Abnova Corporation Supplier No. MAB3951
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Chlamydia species LPS.

Specifications

Antigen Chlamydia species LPS
Applications ELISA, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone BDI168
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Chlamydia species LPS.
Dilution Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native from Chlamydia trachomatis LGV 2 elementary bodies.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity LPS, Genus specific. Reactive with elementary bodies from L2, D, E, F, G, H, I, J, K.
Target Species Bacteria
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.