Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlamydia trachomatis MOMP, Mouse, Clone: CL12-671.11.23, Abnova™

Catalog No. 89136109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-109 Supplier Abnova Corporation Supplier No. MAB6152
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.

Specifications

Antigen Chlamydia trachomatis MOMP
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone CL12-671.11.23
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Chlamydia trachomatis elementary bodies.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies recognize Chlamydia trachomatis serovars A, B, Ba, C, D, E, F, G, H, I, J, K, L1, L2, L3.
Target Species Bacteria
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.