Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum D toxoid, Mouse, Clone: KB22, Abnova™

Catalog No. 89134618 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-618 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-618 Supplier Abnova Corporation Supplier No. MAB0406

Mouse monoclonal antibody raised against Clostridium botulinum D Toxoid.

Specifications

Antigen Clostridium botulinum D toxoid
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone KB22
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Clostridium botulinum D Toxoid.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Clostridium botulinum D toxoid
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to Clostridium botulinum D toxoid. No cross-reaction with Clostridium botulinum A and B toxins.
Target Species Bacteria
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.