Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum Toxin A, Mouse, Clone: B365M, Abnova™

Catalog No. 89135613 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-613 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-613 Supplier Abnova Corporation Supplier No. MAB4095
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A.

Specifications

Antigen Clostridium botulinum Toxin A
Applications ELISA, Immunocytochemistry, Immunofluorescence, Radioimmune Assays (RIA)
Classification Monoclonal
Clone B365M
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A.
Dilution Immunocytochemistry (1:50) The optimal working dilution should be determined by the end user.
Formulation In culture supernatant
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 1280-1292 of Clostridium botulinum Toxin A.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Binds to intact whole C. botulinum Toxin A, amino acids 1280-1292. Has been found to stain Neuro2A cells treated with GT1b ganglioside prior to the addition of C. botulinum Toxin A. Inhibits the entry of C. botulinum Toxin A to Neuro2A cells treated with GT1b ganglioside prior to the addition of the toxin. Cross-reactivity: C. botulinum Toxin A (1280-1292) 100% C. botulinum Toxin A 100% C. botulinum Toxin B 0% C. botulinum Toxin C 0% C. botulinum Toxin E 0% C. botulinum Toxin F (1272-1280) 0%
Target Species Bacteria
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.