Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum Toxin A SNAP-25, Mouse, Clone: B372M, Abnova™

Catalog No. 89135753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-753 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-753 Supplier Abnova Corporation Supplier No. MAB4733
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A SNAP-25.

Specifications

Antigen Clostridium botulinum Toxin A SNAP-25
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone B372M
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Clostridium botulinum Toxin A SNAP-25.
Dilution Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In cell culture supernatant
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 183-197 of Clostridium botulinum Toxin A SNAP-25.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for both the intact whole form of SNAP-25 and the C. botulinum Toxin A cleaved form of SNAP-25. Has been found to bind to cleaved and uncleaved SNAP-25 amino acids 183-197 by ELISA and cell staining techniques. Cross-reactivity: SNAP-25 (cleaved by C. botulinum Toxin A) 100% SNAP-25 (uncleaved by C. botulinum Toxin A) 100% VAMP2 0% Syntaxin 1 0%
Target Species Bacteria
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.