Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corynebacterium diphtheriae, Mouse, Clone: DiD1, Abnova™

Catalog No. 89134925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-925 Supplier Abnova Corporation Supplier No. MAB1292
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Corynebacterium diphtheriae.

Specifications

Antigen Corynebacterium diphtheriae
Applications ELISA, Western Blot
Classification Monoclonal
Clone DiD1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Corynebacterium diphtheriae.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.1% gelatin, 0.09% sodium azide)
Host Species Mouse
Immunogen Supernatant from Corynebacterium diphtheriae.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with epitope of free B subunit and on whole Diphteria toxin molecule and do not react with free A subunit.
Content And Storage Store at 4°C for short term storage. Aliquot and store at -20°C or -80°C.for long term storage.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.