Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corynebacterium diphtheriae Diphteria Toxin, Mouse, Clone: 3B6, Abnova™

Catalog No. 89135258 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-258 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-258 Supplier Abnova Corporation Supplier No. MAB2821
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Corynebacterium diphtheriae Diphteria Toxin.

Specifications

Antigen Corynebacterium diphtheriae Diphteria Toxin
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Corynebacterium diphtheriae Diphteria Toxin.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Corynebacterium diphtheriae Diphteria Toxin.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody does not react with free A or B subunits of Diphteria toxin.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.