Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Cryptosporidium parvum, Clone: BDI370, Abnova™

Catalog No. 89135561 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-561 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-561 Supplier Abnova Corporation Supplier No. MAB3964
Add to Cart
Add to Cart

Specifications

Classification Monoclonal
Clone BDI370
Description Mouse monoclonal antibody raised against Cryptosporidium parvum.
Dilution Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Immunogen Native purified from oocysts.
Quantity 1 mg
Primary or Secondary Primary
Test Specificity Reacts with intact oocysts of C. parvum.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.