Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-Ek, Mouse, Clone: 14-4-4S, Abnova™
Mouse monoclonal antibody raised against native I-Ek.
Supplier: Abnova Corporation MAB3254
Specifications
I-Ek | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
500 μg | |
Primary | |
Mouse | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
14-4-4S | |
Mouse monoclonal antibody raised against native I-Ek. | |
In 100mM BBS, pH 8.2 | |
Native from C3H mouse skin graft and splenocytes. | |
RUO | |
An epitope on mouse I-Ek MHC class II alloantigen. | |
Store at 4°C. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction