Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
I-Ek, Mouse, PE, Clone: 17-3-3S, Abnova™
Mouse monoclonal antibody raised against I-Ek.
Supplier: Abnova Corporation MAB5806
Specifications
I-Ek | |
Monoclonal | |
PE | |
Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Mouse | |
Liquid |
Flow Cytometry | |
17-3-3S | |
Mouse monoclonal antibody raised against I-Ek. | |
In PBS (0.09% sodium azide) | |
Native purified I-Ek from C3H mouse skin graft and splenocytes. | |
RUO | |
An epitope on mouse I-Ek MHC class II alloantigen. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction