Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli, Mouse, Clone: 110, Abnova™

Catalog No. 89135572 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-572 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-572 Supplier Abnova Corporation Supplier No. MAB3981
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Escherichia coli.

Specifications

Antigen Escherichia coli
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Escherichia coli.
Dilution ELISA(1:20-1:200) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Mixture of Escherichia coli serotypes: O18, O112 and O125., O44
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with a number of E. coli serotypes. Reactive with at least the following E. coli serotypes: O18, O44, O112 and O125. It does not cross-react with other members of the Enterobacteriaceae.
Target Species Bacteria, E. coli
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.