Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli, Mouse, Clone: 3011, Abnova™

Catalog No. 89135722 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-722 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-722 Supplier Abnova Corporation Supplier No. MAB4679

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against Escherichia coli.

Specifications

Antigen Escherichia coli
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 3011
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Escherichia coli.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified LPS from Escherichia coli O157.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for E. coli serotype O157 by immunofluorescence. Immunoprecipitates LPS of O157:H7. The region recognized by this antibody is LPS oligosaccharide. Non-reactive with E. coli serotypes: O111, O125, O20, O55 and K12
Target Species E. coli
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.