Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Escherichia coli, Clone: 404, Abnova™

Catalog No. 89135618 Shop All Abnova Corporation Products
Change view
Click to view available options
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-135-618 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-618 Supplier Abnova Corporation Supplier No. MAB4117

Specifications

Classification Monoclonal
Formulation In buffer containing 0.09% sodium azide
Primary or Secondary Primary
Isotype IgG2b Kappa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.