Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli K99 pili, Mouse, Clone: K9912-418.2, Abnova™

Catalog No. 89136464 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-464 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-464 Supplier Abnova Corporation Supplier No. MAB8332
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Escherichia coli K99 pili.

Specifications

Antigen Escherichia coli K99 pili
Applications ELISA
Classification Monoclonal
Clone K9912-418.2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Escherichia coli K99 pili.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Purified Escherichia coli K99 pili.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies do not cross-react with K99-negative E. coli, E. coli K88ac, E. coli 987P, Proteus mirabilis, Proteus vulgaris, Pseudomonas aeruginosa, Enterobacter aerogenes, Citrobacter freundii, Klebsiella pneumoniae, or Strep. agalactiae.
Target Species Bacteria, E. coli
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.