Learn More
Specifications
Specifications
| Antigen | Escherichia coli O157 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Clone | BDI160 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against Escherichia coli O157. |
| Dilution | Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user. |
| Formulation | In 10mM PBS, pH 7.2 (0.09% sodium azide) |
| Host Species | Mouse |
| Immunogen | Native from Escherichia coli. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.