Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Escherichia coli O157, Mouse, FITC, Clone: BDI160, Abnova™

Catalog No. 89135988 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-988 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-988 Supplier Abnova Corporation Supplier No. MAB5484

Mouse monoclonal antibody raised against Escherichia coli O157.

Specifications

Antigen Escherichia coli O157
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone BDI160
Conjugate FITC
Formulation In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Host Species Mouse
Immunogen Escherichia coli O157 whole cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for E. coli serotype O157. Non-reactive with E. coli serotypes O111, O125, O20, O55 and K12.
Target Species E. coli
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.