Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giardia lamblia, Mouse, Clone: BDI612, Abnova™

Catalog No. 89135734 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-734 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-734 Supplier Abnova Corporation Supplier No. MAB4711
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Giardia lamblia.

Specifications

Antigen Giardia lamblia
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone BDI612
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Giardia lamblia.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Intact formalin-fixed cysts.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific to the 65KDa Giardia antigen. Detects cysts.
Target Species Protozoa
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.