Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis E virus ORF 3, Mouse, Clone: 5576f, Abnova™
Mouse monoclonal antibody raised against partial recombinant Hepatitis E virus ORF 3.
Supplier: Abnova Corporation MAB8940
Specifications
Hepatitis E virus ORF 3 | |
Monoclonal | |
1 ug/uL | |
Mouse monoclonal antibody raised against partial recombinant Hepatitis E virus ORF 3. | |
In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
Recombinant protein corresponding to amino acids 34-123 of Hepatitis E virus ORF 3. | |
100 μL | |
Primary | |
Virus | |
Liquid |
ELISA, Western Blot | |
5576f | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity purification | |
RUO | |
This antibody recognizes recombinant protein Hepatitis E virus ORF 3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction