Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Legionella Antibody (LP46), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064479AF405
Description
Legionella Monoclonal specifically detects Legionella in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Legionella | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Native | |
0.1 mL | |
Primary | |
Protein A purified | |
Store at 4C in the dark. | |
IgG3 |
Immunocytochemistry, Immunofluorescence | |
LP46 | |
Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Strongly reactive with SG1 (Pontiac 1, Benidorm, Knoxville). Also reacts with SG1 (Philadelphia). No other serogroups reactive. No reactivity with L. erythra, L. dumoffii, E. coli or Pseudomonas aeruginosa. | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction