Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella pneumophila groEL, Mouse, Clone: 3d566, Abnova™

Catalog No. 89113651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-651 Supplier Abnova Corporation Supplier No. MAB8928
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Legionella pneumophila groEL.

Specifications

Antigen Legionella pneumophila groEL
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3d566
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Legionella pneumophila groEL.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Gene groEL
Gene Symbols groEL
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 72-478 of Legionella pneumophila groEL.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3078335
Test Specificity This antibody recognizes recombinant protein LP HSP60.
Target Species L. pneumophila
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.