Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella pneumophila LPS, Mouse, Clone: 5F4, Abnova™

Catalog No. 89135371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-371 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-371 Supplier Abnova Corporation Supplier No. MAB3159

Mouse monoclonal antibody raised against Legionella Pneumophila LPS.

Specifications

Antigen Legionella pneumophila LPS
Applications ELISA
Classification Monoclonal
Clone 5F4
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Membrane fraction from strain Philadelphia 1 of Legionella pneumophila.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Crocodile
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.