Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Legionella pneumophila mip, Mouse, Clone: 223v5, Abnova™

Catalog No. 89136631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-631 Supplier Abnova Corporation Supplier No. MAB8918
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant Legionella pneumophila mip.

Specifications

Antigen Legionella pneumophila mip
Applications ELISA, Western Blot
Classification Monoclonal
Clone 223v5
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Legionella pneumophila mip.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 33-206 of Legionella pneumophila mip.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes recombinant protein LP mip.
Target Species L. pneumophila
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.