Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis 38kDa, Mouse, Clone: A11D2, Abnova™

Catalog No. 89136422 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-422 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-422 Supplier Abnova Corporation Supplier No. MAB8130
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis 38kDa antigen.

Specifications

Antigen Mycobacterium tuberculosis 38kDa
Applications ELISA, Western Blot
Classification Monoclonal
Clone A11D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis 38Kd antigen.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Host Species Mouse
Immunogen Recombinant protein corresponding to virulent strain H37Rv of Mycobacterium tuberculosis 38Kd antigen.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.