Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis Rv2626, Mouse, Clone: 26A8, Abnova™

Catalog No. 89135395 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-395 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-395 Supplier Abnova Corporation Supplier No. MAB3183

Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis Rv2626.

Specifications

Antigen Mycobacterium tuberculosis Rv2626
Applications ELISA, Western Blot
Classification Monoclonal
Clone 26A8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis Rv2626.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to Mycobacterium tuberculosis Rv2626.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes amino acid sequence corresponding to Rv2626 gene.
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.