Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neisseria gonorrhoeae, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89136142 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-142 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-142 Supplier Abnova Corporation Supplier No. MAB6185
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Neisseria gonorrhoeae.

Specifications

Antigen Neisseria gonorrhoeae
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Neisseria gonorrhoeae.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen 5766 and 8038 (W-II), 8660 (W-III)., Native Neisseria gonorrhoeae cells: Neisseria Reference Laboratory strains G-7, R-11and 7122 (W-I)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies do not cross-react with N. meningitidis, N. cinerea, N. lactamica, N. sicca, B. catarrhalis, E. coli, P. mirabilis, Gardnerella vaginialis, Group B Strep. or Chlamydia trachomatis.
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.