Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neisseria meningitidis GNA2132, Mouse, Clone: 4b1d6, Abnova™

Catalog No. 89136628 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-136-628 Supplier Abnova Corporation Supplier No. MAB8910
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant Neisseria meningitidis GNA2132.

Specifications

Antigen Neisseria meningitidis GNA2132
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4b1d6
Concentration 1 ug/uL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant Neisseria meningitidis GNA2132.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300)
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 86-282 of Neisseria meningitidis GNA2132.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes recombinant protein NM GNA2132.
Target Species Bacteria
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.