Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas aeruginosa serotype 16C, Mouse, Clone: 6008, Abnova™

Catalog No. 89135183 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-183 Supplier Abnova Corporation Supplier No. MAB2433
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Pseudomonas aeruginosa serotype 16C.

Specifications

Antigen Pseudomonas aeruginosa serotype 16C
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 6008
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Pseudomonas aeruginosa serotype 16C.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified Pseudomonas aeruginosa serotype 16C.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with P. aeruginosa serotype 16c.
Target Species P. aeruginosa
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.