Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Salmonella paratyphi A, Mouse, Clone: 10B10G, Abnova™

Catalog No. 89136359 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-359 Supplier Abnova Corporation Supplier No. MAB7805
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Salmonella paratyphi A.

Specifications

Antigen Salmonella paratyphi A
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 10B10G
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Salmonella paratyphi A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native Salmonella paratyphi.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody detects Salmonella paratyphi. It does not react with other Salmonella, E. coli, or Klebsiella.
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.