Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus aureus, Mouse, Clone: 704, Abnova™

Catalog No. 89135706 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-706 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-706 Supplier Abnova Corporation Supplier No. MAB4340

Mouse monoclonal antibody raised against Staphylococcus aureus.

Specifications

Antigen Staphylococcus aureus
Applications ELISA
Classification Monoclonal
Clone 704
Conjugate Unconjugated
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native Staphylococcus aureus (ATCC #29740).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with Staphylococcus aureus, Protein A-negative Staphylococcus aureus and Staphylococcus epidermidis. Does not cross-react with Corynebacteria species, Streptococcus species or Mycoplasma species.
Target Species Staphylococcus species
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.