Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Staphylococcus aureus enterotoxin A from FRI-722H strain, Mouse, Clone: S2, Abnova™
Mouse monoclonal antibody raised against enterotoxin A from FRI-722H strain of Staphylococcus aureus.
Supplier: Abnova Corporation MAB0437
Specifications
Staphylococcus aureus enterotoxin A from FRI-722H strain | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
200 μg | |
Primary | |
Store at 4°C. | |
IgG2a |
ELISA | |
S2 | |
Mouse monoclonal antibody raised against enterotoxin A from FRI-722H strain of Staphylococcus aureus. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
Enterotoxin A from FRI-722H strain of Staphylococcus aureus. | |
RUO | |
Bacteria, Staphylococcus species | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction