Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus aureus enterotoxin A from FRI-722H strain, Mouse, Clone: S5, Abnova™

Catalog No. 89134635 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-635 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-635 Supplier Abnova Corporation Supplier No. MAB0438

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against enterotoxin A from FRI-722H strain of Staphylococcus aureus.

Specifications

Antigen Staphylococcus aureus enterotoxin A from FRI-722H strain
Applications ELISA
Classification Monoclonal
Clone S5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against enterotoxin A from FRI-722H strain of Staphylococcus aureus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Enterotoxin A from FRI-722H strain of Staphylococcus aureus.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria, Staphylococcus species
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.