Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staphylococcus aureus enterotoxin B, Mouse, Clone: S643, Abnova™

Catalog No. 89134632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-632 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-632 Supplier Abnova Corporation Supplier No. MAB0435
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant enterotoxin B of Staphylococcus aureus.

Specifications

Antigen Staphylococcus aureus enterotoxin B
Applications ELISA
Classification Monoclonal
Clone S643
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant enterotoxin B of Staphylococcus aureus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Recombinant protein corresponding to enterotoxin B of Staphylococcus aureus.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to enterotoxin B of Staphylococcus aureus, do not cross-react with St. aureus enterotoxins A, C, D and E.
Target Species Bacteria
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.