Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus agalactiae, Mouse, Clone: Strep12-504.5, Abnova™

Catalog No. 89136486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-486 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-486 Supplier Abnova Corporation Supplier No. MAB8390
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Streptococcus agalactiae.

Specifications

Antigen Streptococcus agalactiae
Applications ELISA
Classification Monoclonal
Clone Strep12-504.5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Streptococcus agalactiae.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Host Species Mouse
Immunogen Native Streptococcus agalactiae cells isolated from mastitic milk.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity These antibodies recognize Strep. agalactiae and cross-react weakly with Strep. dysgalactiae. They do not crossreact with Strep. uberis, Strep. bovis, Strep. faecalis, Staph. aureus, Mycoplasma bovis, Mycoplasma californicum, Mycoplasma bovigenitalium, E. coli, or Corynebacterium bovis.
Target Species Bacteria
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.