Learn More
Mouse anti-Streptococcus Group A (carbohydrate), FITC, Clone: 3401, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Classification | Monoclonal |
| Conjugate | FITC |
| Formulation | In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide) |
| Primary or Secondary | Primary |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.