Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus Group A Mouse, Clone: 501, Abnova™

Catalog No. 89135708 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-708 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-708 Supplier Abnova Corporation Supplier No. MAB4342
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Streptococcus Group A.

Specifications

Antigen Streptococcus Group A
Applications ELISA
Classification Monoclonal
Clone 501
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Streptococcus Group A.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Host Species Mouse
Immunogen Native Streptococcus agalactiae.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes Streptococcus Group A. Does not cross react with Streptococcus groups B-G, Streptococcus pneumonia, Staphylococcus aureus, Corynebacterium dyptheriae, Streptococcus salivarious or Micrococcus mucilagines.
Target Species Streptococcus species
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.