Learn More
ICAM1, Mouse, Clone: 13, Abnova™
Mouse monoclonal antibody raised against partial recombinant ICAM1.
Supplier: Abnova Corporation MAB9880
Description
This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor. [provided by RefSeq
Specifications
ICAM1 | |
Monoclonal | |
Unconjugated | |
Western blot (1:1000) The optimal working dilution should be determined by the end user. | |
ICAM1 | |
BB2/CD54/P3.58 | |
Mouse | |
Affinity purification | |
RUO | |
3383 | |
Human | |
Liquid |
ELISA, Western Blot | |
13 | |
Mouse monoclonal antibody raised against partial recombinant ICAM1. | |
In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300) | |
NM_000201.2 | |
ICAM1 | |
Recombinant protein corresponding to amino acids 301-532 of human ICAM1. | |
100 μg | |
Primary | |
It can expression in SW480 whole cell lysate. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.