Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM1, Mouse, Clone: MEM 112, Abnova™

Catalog No. 89112207 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-207 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-207 Supplier Abnova Corporation Supplier No. MAB5292
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ICAM1.

This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor. [provided by RefSeq

Specifications

Antigen ICAM1
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone MEM 112
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ICAM1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene ICAM1
Gene Alias BB2/CD54/P3.58
Gene Symbols ICAM1
Host Species Mouse
Immunogen Native purified ICAM1 from Raji cell line.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3383
Test Specificity This antibody reacts with human broadly expressed adhesive molecule ICAM-1, the 90 kD glycoprotein Intracellular Adhesion Molecule-1 (CD54). The adhesion molecule ICAM-1 belongs to the immunoglobulin family, it is a ligand for the intergrin LFA-1.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.