Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM1, Mouse, FITC, Clone: 1H4, Abnova™

Catalog No. 89112105 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-105 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-105 Supplier Abnova Corporation Supplier No. MAB5087
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ICAM1.

This gene encodes a cell surface glycoprotein which is typically expressed on endothelial cells and cells of the immune system. It binds to integrins of type CD11a / CD18, or CD11b / CD18 and is also exploited by Rhinovirus as a receptor. [provided by RefSeq

Specifications

Antigen ICAM1
Applications Flow Cytometry
Classification Monoclonal
Clone 1H4
Conjugate FITC
Description Mouse monoclonal antibody raised against native ICAM1.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene ICAM1
Gene Alias BB2/CD54/P3.58
Gene Symbols ICAM1
Host Species Mouse
Immunogen Native purified ICAM1 from Raji cells and spleen cells fused with NS1 cells.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3383
Test Specificity This antibody reacts with CD54 (ICAM-1), a 85-110 KDa type I transmembrane glycoprotein (receptor for rhinovirus) expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells; the expression of CD54 is upregulated by activation.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.